tert-Butyl-5-bromoindoline-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Boc-5-bromoindoline, tert-Butyl 5-bromoindoline-1-carboxylate
H59341-03
H59341-MD
CAAAH59341-03EA
333.38
CAD
CAAAH59341-03
CAAAH59341-MD
tert-Butyl-5-bromoindoline-1-carboxylate 97%
tert-Butyl-5-bromoindoline-1-carboxylate
Purity: 97%
CAS No.: 261732-38-1
Molecular Formula: C13H16BrNO2
Formula Weight: 298.18
MDL No.: MFCD08059280
Formula:
C13H16BrNO2 MW: 298.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08059280 CAS Number: 261732-38-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...