1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyanopyridinium agent, 1-Cyano-4-(dimethylamino)pyridin-1-ium tetrafluoroborate, CDAP, 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate

H59334-MC H59334-ME
CAAAH59334-MCEA 180.12 CAD
CAAAH59334-MC CAAAH59334-ME
1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate 98%
1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate

Purity: 98% CAS No.: 59016-56-7 Molecular Formula: C8H10BF4N3 Formula Weight: 234.99 Storage Temperature: -30°C to -10°C Melting Point: 196-200° Beilstein Reference: 5685616 MDL No.: MFCD00011998

Formula: C₈H₁₀BF₄N₃
MW: 234.99 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011998
CAS Number: 59016-56-7
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR