Azetidin-3-amine dihydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Aminoazetidine dihydrochloride
H59318-MD
H59318-ME
CAAAH59318-MDEA
259.12
CAD
CAAAH59318-MD
CAAAH59318-ME
Azetidin-3-amine dihydrochloride 95%
Azetidin-3-amine dihydrochloride
Purity: 95%
CAS No.: 102065-86-1
Molecular Formula: C3H8N2.2HCl
Formula Weight: 145.03
MDL No.: MFCD09910173
Formula:
C₃H₁₀Cl₂N₂ MW: 145.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09910173 CAS Number: 102065-89-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...