Fmoc-Cys(Acm)-OH 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59306-03
H59306-06
CAAAH59306-03EA
39.34
CAD
CAAAH59306-03
CAAAH59306-06
Fmoc-Cys(Acm)-OH 95%
Fmoc-Cys(Acm)-OH
Purity: 95%
CAS No.: 86060-81-3
Molecular Formula: C21H22N2O5S
Formula Weight: 414.47
Storage Temperature: 2°C to 8°C
Melting Point: 147°
Beilstein Reference: 4725388
Optical Rotation: -33° (c=1 in methanol)
MDL No.: MFCD00038769
Formula:
C₂₁H₂₂N₂O₅S Melting Pt: 147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038769 CAS Number: 86060-81-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...