Hexadecyl palmitate 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate
Palmityl palmitate is used as emollients, masking agents, and skin conditioning. Used in skin and hair applications, it improves emulsion texture and stability and gives structure to cosmetic sticks.
- Useful as glosser and thickener for creams
- Palmityl palmitate improves emulsion texture and stability
Purity: 96%
CAS No.: 540-10-3
Molecular Formula: C32H62O2
Formula Weight: 480.85
Storage Temperature: 2°C to 8°C
Melting Point: 55-56°
MDL No.: MFCD00053739
Formula:
C₃₂H₆₄O₂ MW: 480.86 g/mol Melting Pt: 55…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053739 CAS Number: 540-10-3 EINECS: 208-736-6 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...