Butyl levulinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Levulinic acid butyl ester, 4-Oxovaleric acid butyl ester, Butyl 4-oxovalerate

H59274-AE H59274-AP
CAAAH59274-AEEA 34.44 CAD
CAAAH59274-AE CAAAH59274-AP
Butyl levulinate 98%
Butyl levulinate

CAS: 2052-15-5 MDL: MFCD00009449

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
Boiling Pt: 106
Density: 0.974
Flash Pt: 92 °C(198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009449
CAS Number: 2052-15-5
EINECS: 218-143-4

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR