4-Nitrothiophenol 96%

Supplier: Thermo Scientific Chemicals
Warning

H59240-06 H59240-14
CAAAH59240-06EA 162.74 CAD
CAAAH59240-06 CAAAH59240-14
4-Nitrothiophenol 96%
4-Nitrothiophenol

CAS: 1849-36-1 MDL: MFCD00007343

Formula: C₆H₅NO₂S
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00007343
CAS Number: 1849-36-1
EINECS: 217-436-4

Order Now

Specification Test Results

Assay Minimum 80


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR