1-Ethyl-2,3-dimethylimidazolium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59225-06
H59225-18
CAAAH59225-06EA
126.87
CAD
CAAAH59225-06
CAAAH59225-18
1-Ethyl-2,3-dimethylimidazolium chloride 97%
1-Ethyl-2,3-dimethylimidazolium chloride
CAS: 92507-97-6
MDL: MFCD03093290
Formula:
C₇H₁₃ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD03093290 CAS Number: 92507-97-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...