3-Bromo-5-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H59224-03 H59224-06
CAAAH59224-03EA 65.57 CAD
CAAAH59224-03 CAAAH59224-06
3-Bromo-5-formylpyridine 97%
3-Bromo-5-formylpyridine

CAS: 113118-81-3 MDL: MFCD03265758

Formula: C₆H₄BrNO
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR