4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

H59222-03 H59222-MD
CAAAH59222-03EA 276.5 CAD
CAAAH59222-03 CAAAH59222-MD
4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester 95%
4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 775351-40-1 Molecular Formula: C13H17BN4O2 Formula Weight: 272.11 MDL No.: MFCD12761127

Formula: C₁₃H₁₇BN₄O₂
Storage Temperature: Ambient
MDL Number: MFCD12761127
CAS Number: 775351-40-1

Order Now

Specification Test Results

Assay Minimum 95

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR