5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98% (by HPLC)
Supplier: Thermo Scientific Chemicals
H59169-06
H59169-14
CAAAH59169-06EA
97.64
CAD
CAAAH59169-06
CAAAH59169-14
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98% (by HPLC)
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate
Purity: 98%
CAS No.: 71989-18-9
Molecular Formula: C24H27NO6
Formula Weight: 425.48
Storage Temperature: 2°C to 8°C
Melting Point: 83-90°
Beilstein Reference: 3636375
Optical Rotation: -5.0° (c=1 in acetic acid/water (4:1))
MDL No.: MFCD00037135
Formula:
C₂₄H₂₇NO₆ Melting Pt: 83…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037135 CAS Number: 71989-18-9 EINECS: 276-253-8 |
Specification Test Results
Assay (HPLC) | Min. 97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...