N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%
Supplier: Thermo Scientific Chemicals
H59163-06
H59163-14
CAAAH59163-06EA
274.92
CAD
CAAAH59163-06
CAAAH59163-14
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt
Purity: 98%
CAS No.: 18942-50-2
Molecular Formula: C24H46N2O5
Formula Weight: 442.63
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 161-164°
Beilstein Reference: 5207271
Optical Rotation: +25° (c=1 in DMF)
MDL No.: MFCD00065590
Formula:
C₂₄H₄₆N₂O₅ Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065590 CAS Number: 18942-50-2 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...