2-Amino-4-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

H59130-06 H59130-14
CAAAH59130-06EA 94.8 CAD
CAAAH59130-06 CAAAH59130-14
2-Amino-4-chloropyridine 97%
2-Amino-4-chloropyridine

CAS: 19798-80-2 MDL: MFCD04113820

Formula: C₅H₅ClN₂
MW: 128.56
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR