L-Phenylalanine-tert-butyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe-OtBu·HCl, H-Phe-OtBu.HCl

H59122-03 H59122-06
CAAAH59122-03EA 91.64 CAD
CAAAH59122-03 CAAAH59122-06
L-Phenylalanine-tert-butyl ester hydrochloride 98%
L-Phenylalanine-tert-butyl ester hydrochloride

Purity: 98% CAS No.: 15100-75-1 Molecular Formula: C13H19NO2.HCl Formula Weight: 257.76 Storage Temperature: 2°C to 8°C Melting Point: 240° Beilstein Reference: 4723424 Optical Rotation: +47° (c=2 in ethanol) MDL No.: MFCD00038894

Formula: C₁₃H₂₀ClNO₂
MW: 257.758 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00038894
CAS Number: 15100-75-1
EINECS: 239-151-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR