Nα-Fmoc-N-δ-trityl-L-glutamine 98%

Supplier: Thermo Scientific Chemicals
Warning

H59080-06 H59080-14
CAAAH59080-06EA 102.54 CAD
CAAAH59080-06 CAAAH59080-14
Nα-Fmoc-N-δ-trityl-L-glutamine 98%
N-alpha-Fmoc-N-delta-trityl-L-glutamine

Purity: 98% CAS No.: 132327-80-1 Molecular Formula: C39H34N2O5 Formula Weight: 610.71 Storage Temperature: 2°C to 8°C Melting Point: 165-172° Beilstein Reference: 4343953 Optical Rotation: -14° (c=1 in DMF) MDL No.: MFCD00077056

Formula: C₃₉H₃₄N₂O₅
Melting Pt: 165…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00077056
CAS Number: 132327-80-1

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR