4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropane-1-carbonitrile, 1-[3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropanecarbonitrile, 4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester

H59075-03 H59075-MD
CAAAH59075-03EA 850.04 CAD
CAAAH59075-03 CAAAH59075-MD
4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester 96%
4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C16H19BFNO2 Formula Weight: 287.14 MDL No.: MFCD22418790

Formula: C₁₆H₁₉BFNO₂
MW: 287.14 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD22418790
CAS Number: 917397-94-5

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR