3-Benzyloxy-2-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

H59049-03 H59049-MD
CAAAH59049-03EA 671.5 CAD
CAAAH59049-03 CAAAH59049-MD
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester 96%
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester

MDL: MFCD22419264

Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD22419264
CAS Number: 874152-84-8

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR