Fmoc-Arg(Pbf)-OH 98%
Supplier: Thermo Scientific Chemicals
H59039-06
H59039-14
CAAAH59039-06EA
114.71
CAD
CAAAH59039-06
CAAAH59039-14
Fmoc-Arg(Pbf)-OH 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine
Purity: 98%
CAS No.: 154445-77-9
Molecular Formula: C34H40N4O7S
Formula Weight: 648.77
Storage Temperature: -30°C to -10°C
Beilstein Reference: 8302671
Optical Rotation: -5.5° (c=1 in DMF)
MDL No.: MFCD00235804
Formula:
C₃₄H₄₀N₄O₇S Storage Temperature: Refrigerator |
MDL Number:
MFCD00235804 CAS Number: 154445-77-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...