3-Amino-2,4-difluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Amino-2,4-difluorophenylboronic acid pinacol ester

H59033-03 H59033-MD
CAAAH59033-03EA 715.74 CAD
CAAAH59033-03 CAAAH59033-MD
3-Amino-2,4-difluorobenzeneboronic acid pinacol ester 96%
3-Amino-2,4-difluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C12H16BF2NO2 Formula Weight: 255.07 Melting Point: 75-77° MDL No.: MFCD22419270

Formula: C₁₂H₁₆BF₂NO₂
MW: 255.07 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD22419270
CAS Number: 1315692-91-1

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR