N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59027-03
H59027-06
CAAAH59027-03EA
53.56
CAD
CAAAH59027-03
CAAAH59027-06
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Purity: 97%
CAS No.: 74844-91-0
Molecular Formula: C11H19NO5
Formula Weight: 245.28
Melting Point: 92-96°
Optical Rotation: -65° (c=1 in chloroform)
MDL No.: MFCD00076981
Formula:
C₁₁H₁₉NO₅ Melting Pt: 92…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076981 CAS Number: 74844-91-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...