2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,2'-(2,5-Difluoro-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 2,5-Difluorophenyl-1,4-diboronic acid bis(pinacol) ester
H59025-03
H59025-MD
CAAAH59025-03EA
540.36
CAD
CAAAH59025-03
CAAAH59025-MD
2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester 96%
2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester
Purity: 96%
CAS No.: 303006-90-8
Molecular Formula: C18H26B2F2O4
Formula Weight: 366.02
Melting Point: 202-205°
MDL No.: MFCD12407211
Formula:
C₁₈H₂₆B₂F₂O₄ MW: 366.02 g/mol Melting Pt: 202…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD12407211 CAS Number: 303006-90-8 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...