3-Oxetanone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxetan-3-one

H57870-03 H57870-06
CAAAH57870-03EA 225.94 CAD
CAAAH57870-03 CAAAH57870-06
3-Oxetanone 95%
3-Oxetanone

CAS: 6704-31-0 MDL: MFCD09263255

Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 140 °C
Density: 1.124
Flash Pt: 26 °C(79 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD09263255
CAS Number: 6704-31-0
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR