Azetidin-3-ylmethanol hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57801-MD
H57801-ME
CAAAH57801-MDEA
141.25
CAD
CAAAH57801-MD
CAAAH57801-ME
Azetidin-3-ylmethanol hydrochloride 95%
Azetidin-3-ylmethanol hydrochloride
Purity: 95%
CAS No.: 928038-44-2
Molecular Formula: C4H9NO.HCl
Formula Weight: 123.58
MDL No.: MFCD08703352
Formula:
C4H10ClNO Storage Temperature: Ambient |
MDL Number:
MFCD08703352 CAS Number: 928038-44-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...