3-Methoxyazetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H57769-MD H57769-ME
CAAAH57769-MDEA 96.22 CAD
CAAAH57769-MD CAAAH57769-ME
3-Methoxyazetidine hydrochloride 95%
3-Methoxyazetidine hydrochloride

Purity: 95% CAS No.: 148644-09-1 Molecular Formula: C4H9NO.HCl Formula Weight: 123.58 Boiling Point: 143-149° MDL No.: MFCD06804514

Formula: C4H10ClNO
Boiling Pt: 143…149 °C
Storage Temperature: Ambient
MDL Number: MFCD06804514
CAS Number: 148644-09-1

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR