3-Amino-3-methyloxetane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methyloxetan-3-amine, 3-Methyl-3-oxetanamine
H57588-MD
H57588-ME
CAAAH57588-MDEA
304.94
CAD
CAAAH57588-MD
CAAAH57588-ME
3-Amino-3-methyloxetane 95%
3-Amino-3-methyloxetane
Purity: 95%
CAS No.: 874473-14-0
Molecular Formula: C4H9NO
Formula Weight: 87.12
Physical Form: Crystalline solid
Density: 0.99
Boiling Point: 102°
MDL No.: MFCD11111713
Formula:
C₄H₉NO MW: 87.12 g/mol Boiling Pt: 102 °C Density: 0.99 Storage Temperature: Ambient |
MDL Number:
MFCD11111713 CAS Number: 874473-14-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...