1-Boc-3-(bromomethyl)azetidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H57492-MD H57492-ME
CAAAH57492-MDEA 165.9 CAD
CAAAH57492-MD CAAAH57492-ME
1-Boc-3-(bromomethyl)azetidine 95%
1-Boc-3-(bromomethyl)azetidine

Purity: 95% CAS No.: 253176-93-1 Molecular Formula: C9H16BrNO2 Formula Weight: 250.13 MDL No.: MFCD16556174

Formula: C₉H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD16556174
CAS Number: 253176-93-1

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR