4-(trans-4-Amylcyclohexyl)benzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(trans-4-Pentylcyclohexyl)benzoic Acid
H57351-03
H57351-06
CAAAH57351-03EA
64.15
CAD
CAAAH57351-03
CAAAH57351-06
4-(trans-4-Amylcyclohexyl)benzoic acid 99%
4-(trans-4-Amylcyclohexyl)benzoic acid
Purity: 99%
CAS No.: 65355-30-8
Molecular Formula: C18H26O2
Formula Weight: 274.40
MDL No.: MFCD06208360
Formula:
C₁₈H₂₆O₂ MW: 274.40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06208360 CAS Number: 65355-30-8 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...