3-Iodooxetane 95%

Supplier: Thermo Scientific Chemicals
Warning

H57302-MD H57302-ME
CAAAH57302-MDEA 317.58 CAD
CAAAH57302-MD CAAAH57302-ME
3-Iodooxetane 95%
3-Iodooxetane

CAS: 26272-85-5 MDL: MFCD08544403

Formula: C₃H₅IO
Boiling Pt: 166 °C
Density: 2.14
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD08544403
CAS Number: 26272-85-5
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR