5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Benzyl Ester, N-Boc-L-glutamic Acid 5-Benzyl Ester, 5-Benzyl N-Boc-L-glutamate, Boc-Glu(OBzl)-OH
H56879-06
H56879-14
CAAAH56879-06EA
46.93
CAD
CAAAH56879-06
CAAAH56879-14
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate
Purity: 98%
CAS No.: 13574-13-5
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Storage Temperature: 2°C to 8°C
Melting Point: 69-71°
Beilstein Reference: 2226572
Optical Rotation: -5.5° (c=1 in acetic acid)
MDL No.: MFCD00065569
Formula:
C₁₇H₂₃NO₆ MW: 337.37 g/mol Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065569 CAS Number: 13574-13-5 EINECS: 237-007-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...