Benzyl-2-bromoethyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56872-06
H56872-14
CAAAH56872-03EA
72.19
CAD
CAAAH56872-03
CAAAH56872-06
CAAAH56872-14
Benzyl-2-bromoethyl ether 97%
Benzyl-2-bromoethyl ether
Purity: 97%
CAS No.: 1462-37-9
Molecular Formula: C9H11BrO
Formula Weight: 215.09
Flash Point:
Formula:
C₉H₁₁BrO Boiling Pt: 254 °C Density: 1.36 Flash Pt: >113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01321307 CAS Number: 1462-37-9 UN: 3334 ADR: "9", |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...