(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid sesquihydrate, Methyldopa sesquihydrate

H56704-03 H56704-06
CAAAH56704-03EA 31.6 CAD
CAAAH56704-03 CAAAH56704-06
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate

Purity: 99% CAS No.: 41372-08-1 Molecular Formula: C10H13NO4.1.5H2O Formula Weight: 238.24 (211.22anhy) Melting Point: 300° Optical Rotation: -13° (c=1 in water) MDL No.: MFCD00149280

Formula: C₁₀H₁₃NO₄·1.5H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149280
CAS Number: 41372-08-1
Merck Index: 14,6055

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR