2-Naphthalenesulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Naphthalene-2-sulfonic acid, β-Naphthalenesulfonic acid, Naphthalene-2-sulphonic acid, β-Naphthalenesulphonic acid

H56685-22 H56685-36
CAAAH56685-22EA 159.58 CAD
CAAAH56685-22 CAAAH56685-36
2-Naphthalenesulfonic acid 98%
2-Naphthalenesulphonic acid

CAS: 120-18-3 MDL: MFCD00004089

Formula: C₁₀H₈O₃S
Boiling Pt: ~ 350 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.441 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149271
CAS Number: 120-18-3
EINECS: 204-375-3
UN: 2585
ADR: 8,III
Merck Index: 12,06464

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR