D-Pinitol 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-O-Methyl-D-chiro-inositol, (1R,2S,3R,4S,5S,6S)-6-Methoxycyclohexane-1,2,3,4,5-pentaol
H56648-03
H56648-06
H56648-MD
CAAAH56648-03EA
156.1
CAD
CAAAH56648-03
CAAAH56648-06
CAAAH56648-MD
D-Pinitol 95%
D-Pinitol
Purity: 95%
CAS No.: 10284-63-6
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 178-185°
Optical Rotation: +60° (c=1 in water)
MDL No.: MFCD00216659
Formula:
C₇H₁₄O₆ Melting Pt: 178…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216659 CAS Number: 10284-63-6 Merck Index: 14,10361 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...