(+)-Usnic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56612-06
H56612-14
CAAAH56612-06EA
101.75
CAD
CAAAH56612-06
CAAAH56612-14
(+)-Usnic acid 98%
(+)-Usnic acid
Purity: 98%
CAS No.: 7562-61-0
Molecular Formula: C18H16O7
Formula Weight: 344.32
Melting Point: 201-203°
Beilstein Reference: 96698
MDL No.: MFCD00016878
Formula:
C₁₈H₁₆O₇ Melting Pt: 201…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016878 CAS Number: 7562-61-0 EINECS: 231-456-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...