2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,3-Dimercaptopropanesulfonic acid sodium salt, DL-2,3-Dimercapto-1-propanesulphonic acid, sodium salt monohydrate, Sodium 2,3-Dimercaptopropanesulfonate monohydrate, DMPS, Sodium 2,3-dimercaptopropane-1-sulfonate hydrate, Sodium 2,3-dimercaptopropane-1-sulphonate hydrate, DL-2,3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate, 2,3-Dimercaptopropanesulphonic acid sodium salt, Sodium 2,3-Dimercaptopropanesulphonate monohydrate

H56578-03 H56578-MD
CAAAH56578-03EA 344.44 CAD
CAAAH56578-03 CAAAH56578-MD
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate 98%
2,3-Dimercapto-1-propanesulfonic acids odium salt monohydrate

Purity: 98% CAS No.: 207233-91-8 Molecular Formula: C3H7NaO3S3.H2O Formula Weight: 228.30 (210.28anhy) Melting Point: ca 229° dec. Beilstein Reference: 3734863 MDL No.: MFCD00149543

Formula: C₃H₉NaO₄S₃·H₂O
MW: 228.30 g/mol (210.28 anhy)
Melting Pt: ∼ 229 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149543
CAS Number: 207233-91-8
EINECS: 223-796-3
Merck Index: 14,3210

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR