Diethyl cyclopropane-1,1-dicarboxylate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate

H56568-03 H56568-06 H56568-14
CAAAH56568-03EA 54.04 CAD
CAAAH56568-03 CAAAH56568-06 CAAAH56568-14
Diethyl cyclopropane-1,1-dicarboxylate
Diethyl cyclopropane-1,1-dicarboxylate

CAS: 1559-02-0 MDL: MFCD00001283

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Boiling Pt: 94…96 °C/10 mm
Density: 1.055
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR