(S)-(+)-O-Acetylmandelic acid 99%
Supplier: Thermo Scientific Chemicals
H56567-03
H56567-06
CAAAH56567-03EA
49.93
CAD
CAAAH56567-03
CAAAH56567-06
(S)-(+)-O-Acetylmandelic acid 99%
(S)-(+)-O-Acetylmandelic acid
Purity: 99%
CAS No.: 7322-88-5
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 97-100°
Beilstein Reference: 2694109
Optical Rotation: +153° (c=2 in acetone)
MDL No.: MFCD00064215
Formula:
C₁₀H₁₀O₄ Melting Pt: 97…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064215 CAS Number: 7322-88-5 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...