4-Carboxy-1-naphthaleneboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Borono-1-naphthoic acid

H56542-03 H56542-MD
CAAAH56542-03EA 850.04 CAD
CAAAH56542-03 CAAAH56542-MD
4-Carboxy-1-naphthaleneboronic acid 95%
4-Carboxy-1-naphthaleneboronic acid

CAS: 332398-57-9 MDL: MFCD06203538

Formula: C₁₁H₉BO₄
MW: 216.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203538
CAS Number: 332398-57-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR