3,4-Ethylenedioxythiophene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT

H56533-06 H56533-14
CAAAH56533-06EA 80.58 CAD
CAAAH56533-06 CAAAH56533-14
3,4-Ethylenedioxythiophene 97%
3,4-Ethylenedioxythiophene

Often used as a monomer to synthesize the conductive polymer PEDT


Purity: 97% CAS No.: 126213-50-1 Molecular Formula: C6H6O2S Formula Weight: 142.17 Storage Temperature: 2°C to 8°C Flash Point: 104°(219°F) Refractive Index: 1.5765 Boiling Point: 193° UN No.: 2810 MDL No.: MFCD02093622

Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 193 °C
Density: 1.331
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093622
CAS Number: 126213-50-1
EINECS: 603-128-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR