N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97+%

Supplier: Thermo Scientific Chemicals
Danger

H56499-AA H56499-XF
CAAAH56499-AAEA 546.68 CAD
CAAAH56499-AA CAAAH56499-XF
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97+%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane

Purity: 98% CAS No.: 68076-36-8 Molecular Formula: C9H20N2O2 Formula Weight: 188.27 Storage Temperature: 2°C to 8°C Flash Point: 109°(228°F) Refractive Index: 1.4600 Beilstein Reference: 1937878 UN No.: 2735 MDL No.: MFCD00210019

Formula: C₉H₂₀N₂O₂
Density: 0.984
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00210019
CAS Number: 68076-36-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR