(R)-2-Acetoxy-2-phenylacetic acid 98% (by HPLC)
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56492-03
H56492-06
CAAAH56492-03EA
46.45
CAD
CAAAH56492-03
CAAAH56492-06
(R)-2-Acetoxy-2-phenylacetic acid 98% (by HPLC)
(R)-2-Acetoxy-2-phenylacetic acid
The (R)- and (S)-isomers are chiral derivatising agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.
Purity: 98%
CAS No.: 51019-43-3
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 97-99°
Beilstein Reference: 2694110
Optical Rotation: -153° (c=2 in acetone)
MDL No.: MFCD00004249
Caution: Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Formula:
C₁₀H₁₀O₄ Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004249 CAS Number: 51019-43-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...