(S)-(+)-3-Hydroxytetrahydrofuran 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56476-06
H56476-14
CAAAH56476-03EA
41.21
CAD
CAAAH56476-03
CAAAH56476-06
CAAAH56476-14
(S)-(+)-3-Hydroxytetrahydrofuran 98%
(S)-(+)-3-Hydroxytetrahydrofuran
CAS: 86087-23-2
MDL: MFCD00064327
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 80 °C/15 mm Density: 1.103 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064327 CAS Number: 86087-23-2 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...