β-(N-Morpholino)ethanesulfonic acid (MES) hydrate ≥99%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Morpholineethanesulphonic acid hydrate, MES hydrate, 2-(4-Morpholinyl)ethanesulphonic acid hydrate, 4-Morpholineethanesulfonic acid hydrate, 2-(4-Morpholinyl)ethanesulfonic acid hydrate
MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good?s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
Purity: 99+%
CAS No.: 1266615-59-1
Molecular Formula: C6H13NO4S.xH2O
Formula Weight: 195.24(anhy)
Melting Point: ca 308° dec.
Beilstein Reference: 141319
MDL No.: MFCD00149409
Formula:
C₆H₁₃NO₄S^.xH₂O Melting Pt: ca 308 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149409 CAS Number: 1266615-59-1 EINECS: 224-632-3 Merck Index: 14,5902 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...