Azetidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H56351-03
H56351-MD
CAAAH56351-03EA
252.8
CAD
CAAAH56351-03
CAAAH56351-MD
Azetidine 98%
Azetidine
Purity: 98%
CAS No.: 503-29-7
Molecular Formula: C3H7N
Formula Weight: 57.10
Storage Temperature: 2°C to 8°C
Flash Point: -21°(-6°F)
Physical Form: Liquid
Density: 0.847
Refractive Index: 1.4320
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Boiling Point: 61-62°
Beilstein Reference: 102384
UN No.: 2924
MDL No.: MFCD00005165
Formula:
C₃H₇N MW: 57.1 g/mol Boiling Pt: 61…62 °C Density: 0.847 Flash Pt: -21 °C(-6 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005165 CAS Number: 503-29-7 EINECS: 207-963-8 UN: 2733 ADR: "3",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...