Pentacene 97%

Supplier: Thermo Scientific Chemicals

H56310-03 H56310-MD
CAAAH56310-03EA 619.36 CAD
CAAAH56310-03 CAAAH56310-MD
Pentacene 97%
Pentacene

CAS: 135-48-8 MDL: MFCD00003710

Formula: C₂₂H₁₄
Melting Pt: 372…374 °C (subl.)
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 14,7107

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR