Fmoc-L-norvaline 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline
H56290-03
H56290-06
CAAAH56290-03EA
47.08
CAD
CAAAH56290-03
CAAAH56290-06
Fmoc-L-norvaline 98%
Fmoc-L-norvaline
Purity: 98%
CAS No.: 135112-28-6
Molecular Formula: C20H21NO4
Formula Weight: 339.39
Storage Temperature: 2°C to 8°C
Melting Point: 151-155°
Beilstein Reference: 5883879
Optical Rotation: -21° (c=2 in DMF)
MDL No.: MFCD00155631
Formula:
C₂₀H₂₁NO₄ Melting Pt: 151…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155631 CAS Number: 135112-28-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...