4,6-Dichloro-5-nitropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H56252-03 H56252-06 H56252-14
CAAAH56252-03EA 103.65 CAD
CAAAH56252-03 CAAAH56252-06 CAAAH56252-14
4,6-Dichloro-5-nitropyrimidine 98%
4,6-Dichloro-5-nitropyrimidine

CAS: 4316-93-2 MDL: MFCD00006107

Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006107
CAS Number: 4316-93-2
EINECS: 224-340-6

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR