2-Chloro-N-(2,3-dimethylphenyl)benzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H56225-03 H56225-MD
CAAAH56225-03EA 758.4 CAD
CAAAH56225-03 CAAAH56225-MD
2-Chloro-N-(2,3-dimethylphenyl)benzamide 97%
2-Chloro-N-(2,3-dimethylphenyl)benzamide

CAS: 196617-88-6 MDL: MFCD00018254

Formula: C₁₅H₁₄ClNO
MW: 259.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018254
CAS Number: 196617-88-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR