9-Vinylcarbazole ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Vinylcarbazole
H56174-22
H56174-30
CAAAH56174-22EA
436.08
CAD
CAAAH56174-22
CAAAH56174-30
9-Vinylcarbazole ≥98%
9-Vinylcarbazole
Purity: 98+%
CAS No.: 1484-13-5
Molecular Formula: C14H11N
Formula Weight: 193.24
Melting Point: 60-65°
Boiling Point: 154-155°
Beilstein Reference: 132988
UN No.: 2811
MDL No.: MFCD00004966
Formula:
C₁₄H₁₁N Boiling Pt: 154…155 °C Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004966 CAS Number: 1484-13-5 EINECS: 216-055-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...