DL-Cysteine 96%

Supplier: Thermo Scientific Chemicals
Warning

H56126-06 H56126-14
CAAAH56126-06EA 70.78 CAD
CAAAH56126-06 CAAAH56126-14 CAAAH56126-22
DL-Cysteine 96%
DL-Cysteine

CAS: 3374-22-9 MDL: MFCD00004881

Formula: C₃H₇NO₂S
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 225 °C (dec.)
Density: 1.441 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004881
CAS Number: 3374-22-9
EINECS: 222-160-2
Merck Index: 12,02850

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR